ENA423208016 Enamine

[2-(dimethylamino)ethyl][(3-methoxyphenyl)methyl]amine

  • MDL number MFCD07364979
  • Cas Number : 827328-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-168581

Safety Information

msds https://www.enaminestore.com/catalog/EN300-168581
Availability details for
Quantity
  • Availability
  • Add to Cart