ENA423208030 Enamine

[(3-chlorophenyl)methyl](2-methylpropyl)amine

  • MDL number MFCD07407707
  • Cas Number : 893589-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-168595

Safety Information

msds https://www.enaminestore.com/catalog/EN300-168595
Availability details for
Quantity
  • Availability
  • Add to Cart