ENA423208033 Enamine

3-{[(2-methylpropyl)amino]methyl}phenol

  • MDL number MFCD11139768
  • Cas Number : 1019481-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-168598

Safety Information

msds https://www.enaminestore.com/catalog/EN300-168598
Availability details for
Quantity
  • Availability
  • Add to Cart