ENA423208337 Enamine

N-(1-phenylethyl)cyclopentanamine

  • MDL number MFCD03640281
  • Cas Number : 626213-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-168905

Safety Information

msds https://www.enaminestore.com/catalog/EN300-168905
Availability details for
Quantity
  • Availability
  • Add to Cart