ENA423208367 Enamine

N-(propan-2-yl)cycloheptanamine

  • MDL number MFCD08691749
  • Cas Number : 52703-18-1
  • Molecular Weight : 155.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-168935

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-168935
Availability details for
Quantity
  • Availability
  • Add to Cart