ENA423208456 Enamine

N-(3-methoxypropyl)-1-methylpiperidin-4-amine

  • MDL number MFCD01653947
  • Cas Number : 626214-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-169024

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-169024
Availability details for
Quantity
  • Availability
  • Add to Cart