ENA423208922 Enamine

1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]-1,4-diazepane hydrochloride

  • MDL number MFCD27980864
  • Cas Number : 1803607-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-169951

Safety Information

msds https://www.enaminestore.com/catalog/EN300-169951
Availability details for
Quantity
  • Availability
  • Add to Cart