ENA423208934 Enamine

2-(2-amino-3-bromophenyl)propan-2-ol

  • MDL number MFCD27980866
  • Cas Number : 1803605-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170047

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170047
Availability details for
Quantity
  • Availability
  • Add to Cart