ENA423208936 Enamine

4-amino-N-(pyridin-4-ylmethyl)benzamide dihydrochloride

  • MDL number MFCD27959483
  • Cas Number : 1803601-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170056

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170056
Availability details for
Quantity
  • Availability
  • Add to Cart