ENA423208938 Enamine

4-bromo-2-{[(tert-butyldimethylsilyl)oxy]methyl}aniline

  • MDL number MFCD27959485
  • Cas Number : 1214248-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170098

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170098
Availability details for
Quantity
  • Availability
  • Add to Cart