ENA423208940 Enamine

2-ethoxy-4-(4-propylpiperazin-1-yl)aniline trihydrochloride

  • MDL number MFCD27959487
  • Cas Number : 1803611-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170115
Availability details for
Quantity
  • Availability
  • Add to Cart