ENA423208941 Enamine

5-methoxy-4-nitro-1H-indole-3-carboxylic acid

  • MDL number MFCD27980868
  • Cas Number : 135531-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170116
Availability details for
Quantity
  • Availability
  • Add to Cart