ENA423208942 Enamine

5-methoxy-4-nitro-1H-indole-3-carbaldehyde

  • MDL number MFCD27980869
  • Cas Number : 135531-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170118
Availability details for
Quantity
  • Availability
  • Add to Cart