ENA423208950 Enamine

1-[(cyclopentylmethyl)sulfanyl]ethan-1-one

  • MDL number MFCD27980873
  • Cas Number : 1803561-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170261
Availability details for
Quantity
  • Availability
  • Add to Cart