ENA423208951 Enamine

2-bromo-5-(difluoromethyl)-1,3,4-oxadiazole

  • MDL number MFCD27980874
  • Cas Number : 1594041-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. -20

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170289
Availability details for
Quantity
  • Availability
  • Add to Cart