ENA423208957 Enamine

2-chloro-6-(ethanesulfinyl)benzoic acid

  • MDL number MFCD27980877
  • Cas Number : 1803604-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170382
Availability details for
Quantity
  • Availability
  • Add to Cart