ENA423208983 Enamine

tert-butyl 2-(3-amino-2-oxopyrrolidin-1-yl)-3-methylbutanoate

  • MDL number MFCD27980884

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-170728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170728
Availability details for
Quantity
  • Availability
  • Add to Cart