ENA423208986 Enamine

4-chloro-1-(cyclobutylmethoxy)-2-nitrobenzene

  • MDL number MFCD27980887
  • Cas Number : 1803583-46-1
  • Molecular Weight : 241.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-170789

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170789
Availability details for
Quantity
  • Availability
  • Add to Cart