ENA423208987 Enamine

3-(difluoromethoxy)-1,2-oxazole

  • MDL number MFCD27980888
  • Cas Number : 1803562-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170871
Availability details for
Quantity
  • Availability
  • Add to Cart