ENA423208989 Enamine

2-(3,5-dimethylphenyl)pyrrolidine hydrochloride

  • MDL number MFCD27980890
  • Cas Number : 1803595-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170903

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170903
Availability details for
Quantity
  • Availability
  • Add to Cart