ENA423208993 Enamine

3-[(dimethylamino)methyl]benzene-1-carbothioamide hydrochloride

  • MDL number MFCD27980895
  • Cas Number : 1803599-73-6
  • Molecular Weight : 230.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-171105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171105
Availability details for
Quantity
  • Availability
  • Add to Cart