ENA423208995 Enamine

N-(hex-1-yn-3-yl)-2-methylpropane-2-sulfinamide

  • Cas Number : 2168793-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171153

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171153
Availability details for
Quantity
  • Availability
  • Add to Cart