ENA423208999 Enamine

3-methylisoquinoline-5-sulfonamide

  • MDL number MFCD27959521
  • Cas Number : 1823955-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171308
Availability details for
Quantity
  • Availability
  • Add to Cart