ENA423209009 Enamine

4-(2-bromobutyl)pyridine hydrobromide

  • MDL number MFCD27980904
  • Cas Number : 1803584-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171363

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171363
Availability details for
Quantity
  • Availability
  • Add to Cart