ENA423209010 Enamine

8-(9H-fluoren-9-yl)methyl 3-methyl 1-oxa-2,8-diazaspiro[4.5]dec-2-ene-3,8-dicarboxylate

  • MDL number MFCD27980905
  • Cas Number : 1803585-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171364
Availability details for
Quantity
  • Availability
  • Add to Cart