ENA423209011 Enamine

ethyl 5-(furan-3-yl)-1,2-oxazole-4-carboxylate

  • MDL number MFCD27980906
  • Cas Number : 1797582-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171409
Availability details for
Quantity
  • Availability
  • Add to Cart