ENA423209012 Enamine

2-[3-(4-fluoro-3-methylphenyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride

  • MDL number MFCD27980907
  • Cas Number : 1803593-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171531

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171531
Availability details for
Quantity
  • Availability
  • Add to Cart