ENA423209015 Enamine

4-(pyrrolidin-1-yl)pyridine-2-carboximidamide dihydrochloride

  • MDL number MFCD27980910
  • Cas Number : 1803604-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171539

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171539
Availability details for
Quantity
  • Availability
  • Add to Cart