ENA423209399 Enamine

3-amino-N-(cyclopropylmethyl)-4-methylbenzamide

  • MDL number MFCD09729827
  • Cas Number : 953751-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-169906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-169906
Availability details for
Quantity
  • Availability
  • Add to Cart