ENA423209407 Enamine

2-(methoxymethyl)-6-methylmorpholine

  • MDL number MFCD19603230
  • Cas Number : 1339925-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-169930
Availability details for
Quantity
  • Availability
  • Add to Cart