ENA423209417 Enamine

3-(cyclopropylcarbamoyl)benzoic acid

  • MDL number MFCD12173481
  • Cas Number : 852879-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-169973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-169973
Availability details for
Quantity
  • Availability
  • Add to Cart