ENA423209421 Enamine

6-[(cyclopropylmethyl)amino]pyridin-3-ol

  • MDL number MFCD18256586
  • Cas Number : 1256791-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-169987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-169987
Availability details for
Quantity
  • Availability
  • Add to Cart