ENA423209432 Enamine

2-bromo-3,5-difluoroaniline

  • MDL number MFCD16661040
  • Cas Number : 500357-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170009

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170009
Availability details for
Quantity
  • Availability
  • Add to Cart