ENA423209439 Enamine

4-(trifluoromethyl)pyridine-3-sulfonyl chloride

  • MDL number MFCD13189204
  • Cas Number : 533932-27-3
  • Molecular Weight : 245.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(trifluoromethyl)-3-pyridinesulfonyl chloride
Inchi Code 1S/C6H3ClF3NO2S/c7-14(12,13)5-3-11-2-1-4(5)6(8,9)10/h1-3H
Inchi Key VFJOQYGYBJRSJF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-170028

Safety Information

signalword Danger
pcodes P260,P264,P270,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P330,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-170028
pictograms GHS05,GHS07
hstatements H302,H314
Availability details for
Quantity
  • Availability
  • Add to Cart