ENA423209442 Enamine

2-{2-[(2-phenylethyl)amino]ethoxy}ethan-1-ol

  • MDL number MFCD08757924
  • Cas Number : 932278-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-169392

Safety Information

msds https://www.enaminestore.com/catalog/EN300-169392
Availability details for
Quantity
  • Availability
  • Add to Cart