ENA423209450 Enamine

2-bromo-1-(2-bromo-4,5-dimethoxyphenyl)ethan-1-one

  • MDL number MFCD00510456
  • Cas Number : 321681-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170062

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170062
Availability details for
Quantity
  • Availability
  • Add to Cart