ENA423209456 Enamine

6-amino-1-(cyclopentylmethyl)-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD13476188
  • Cas Number : 1308263-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170073

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170073
Availability details for
Quantity
  • Availability
  • Add to Cart