ENA423209457 Enamine

2-ethoxy-4-{octahydropyrrolo[1,2-a]pyrazin-2-yl}aniline

  • MDL number MFCD22895381
  • Cas Number : 1517701-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170080
Availability details for
Quantity
  • Availability
  • Add to Cart