ENA423209461 Enamine

1,2-oxazinane hydrochloride

  • MDL number MFCD13193208
  • Cas Number : 54722-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141-143
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170086
Availability details for
Quantity
  • Availability
  • Add to Cart