ENA423209469 Enamine

2-phenyl-2-[(propan-2-yl)amino]acetic acid

  • MDL number MFCD12048366
  • Cas Number : 557090-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-170102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170102
Availability details for
Quantity
  • Availability
  • Add to Cart