ENA423209473 Enamine

2-bromo-4-nitro-1-(trifluoromethyl)benzene

  • MDL number MFCD16606317
  • Cas Number : 875238-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-170119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170119
Availability details for
Quantity
  • Availability
  • Add to Cart