ENA423209481 Enamine

2-chloro-3-(propan-2-yl)pyrazine

  • MDL number MFCD14702662
  • Cas Number : 57674-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170166

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170166
Availability details for
Quantity
  • Availability
  • Add to Cart