ENA423209486 Enamine

2-[(2-methoxyethoxy)methyl]oxirane

  • MDL number MFCD09802389
  • Cas Number : 13483-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170175

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170175
Availability details for
Quantity
  • Availability
  • Add to Cart