ENA423209490 Enamine

4-methylcycloheptan-1-amine

  • MDL number MFCD19206728
  • Cas Number : 855590-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170185

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170185
Availability details for
Quantity
  • Availability
  • Add to Cart