ENA423209494 Enamine

sodium 2-(trifluoromethyl)benzene-1-sulfinate

  • MDL number MFCD11618069
  • Cas Number : 1009628-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170190
Availability details for
Quantity
  • Availability
  • Add to Cart