ENA423209499 Enamine

1-(2,6-difluorophenyl)-5-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD27980870
  • Cas Number : 1797933-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170198

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170198
Availability details for
Quantity
  • Availability
  • Add to Cart