ENA423209501 Enamine

2-chloro-1-(5-methyl-1,2-oxazol-4-yl)ethan-1-one

  • MDL number MFCD23844909
  • Cas Number : 1517771-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170206

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170206
Availability details for
Quantity
  • Availability
  • Add to Cart