ENA423209505 Enamine

methyl 3-(chlorosulfonyl)-4-methylthiophene-2-carboxylate

  • MDL number MFCD20133310
  • Cas Number : 190375-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170217

Safety Information

msds https://www.enaminestore.com/catalog/EN300-170217
Availability details for
Quantity
  • Availability
  • Add to Cart