ENA423209508 Enamine

1-oxa-3,7-diazaspiro[4.5]decan-2-one hydrochloride

  • MDL number MFCD20278311
  • Cas Number : 1609395-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170229

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170229
Availability details for
Quantity
  • Availability
  • Add to Cart