ENA423209511 Enamine

6-methoxy-5-nitropyrimidine-4-thiol

  • MDL number MFCD09746185
  • Cas Number : 7154-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170247
Availability details for
Quantity
  • Availability
  • Add to Cart